Assay Details
| Gene Symbol: AGL23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105210.2 | NP_176715.1 | 1 - 2 | 314 | 114 | |
| GenBank mRNA | AY141230.1 | - | 1 - 2 | 314 | 114 | |
More Information
Set Membership: |
|

AGL23
Genomic Map