Assay Details
| Gene Symbol: GSTF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111189.3 | NP_186969.1 | 1 - 2 | 325 | 111 | |
| GenBank mRNA | AY099776.1 | - | 1 - 2 | 189 | 111 | |
| AY128877.1 | - | 1 - 2 | 146 | 111 | ||
| U70672.1 | - | 1 - 2 | 151 | 111 | ||
More Information
Set Membership: |
|

GSTF11
Genomic Map