Assay Details
| Gene Symbol: PAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114985.3 | NP_190694.1 | 2 - 3 | 741 | 113 | |
| GenBank mRNA | AF043522.1 | - | 2 - 3 | 595 | 113 | |
| AY042820.1 | - | 2 - 3 | 656 | 113 | ||
| AY052321.1 | - | 2 - 3 | 603 | 113 | ||
| AY061899.1 | - | 2 - 3 | 581 | 113 | ||
| AY081448.1 | - | 2 - 3 | 581 | 113 | ||
| X66825.1 | - | 596 | 113 | |||
More Information
Set Membership: |
|

PAD1
Genomic Map