Assay Details
| Gene Symbol: RABA4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124170.3 | NP_199607.1 | 1 - 2 | 288 | 130 | |
| GenBank mRNA | AF218121.1 | - | 1 - 2 | 288 | 130 | |
| BT024842.1 | - | 1 - 2 | 228 | 130 | ||
More Information
Set Membership: |
|

RABA4C
Genomic Map