Assay Details
| Gene Symbol: PUM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336202.1 | NP_001324126.1 | 2 - 3 | 2457 | 159 | |
| NM_128471.4 | NP_180478.1 | 3 - 4 | 1993 | 159 | ||
More Information
Set Membership: |
|

PUM3
Genomic Map