Assay Details
| Gene Symbol: PUM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336202.1 | NP_001324126.1 | 4 - 5 | 2711 | 150 | |
| NM_128471.4 | NP_180478.1 | 5 - 6 | 2247 | 150 | ||
More Information
Set Membership: |
|

PUM3
Genomic Map