Assay Details
| Gene Symbol: CCDC58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083389.2 | NP_001076858.1 | 4 - 5 | 389 | 109 | |
| GenBank mRNA | BC114911.1 | - | 4 - 5 | 390 | 109 | |
More Information
Set Membership: |
|

CCDC58
Genomic Map