Assay Details
| Gene Symbol: PAFAH1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174663.2 | NP_777088.1 | 7 - 8 | 1515 | 88 | |
| GenBank mRNA | BC140633.1 | - | 7 - 8 | 935 | 88 | |
| D30615.1 | - | 7 - 8 | 1515 | 88 | ||
More Information
Set Membership: |
|

PAFAH1B1
Genomic Map