Assay Details
| Gene Symbol: LRRC57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031754.3 | NP_001026924.2 | 2 - 3 | 201 | 78 | |
| GenBank mRNA | BC118442.1 | - | 2 - 3 | 202 | 78 | |
| BT021726.1 | - | 2 - 3 | 307 | 78 | ||
More Information
Set Membership: |
|

LRRC57
Genomic Map