Assay Details
| Gene Symbol: SELENOT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103103.2 | NP_001096573.2 | 2 - 3 | 367 | 120 | |
| NR_146805.1 | 3 - 4 | 466 | 120 | |||
| GenBank mRNA | BC149082.1 | - | 2 - 3 | 361 | 120 | |
| DT887648.1 | - | 2 - 3 | 183 | 120 | ||
| DV816938.1 | - | 2 - 3 | 298 | 120 | ||
| DV833984.1 | - | 3 - 4 | 368 | 120 | ||
More Information
Set Membership: |
|

SELENOT
Genomic Map