Assay Details
| Gene Symbol: MEAF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024572.3 | NP_001019743.2 | 4 - 5 | 365 | 58 | |
| GenBank mRNA | BC146126.1 | - | 4 - 5 | 366 | 58 | |
| BT021857.1 | - | 4 - 5 | 531 | 58 | ||
More Information
Set Membership: |
|

MEAF6
Genomic Map