Assay Details
| Gene Symbol: HNRNPA2B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045975.1 | NP_001039440.1 | 11 - 12 | 1317 | 132 | |
| GenBank mRNA | BC113298.1 | - | 11 - 12 | 1317 | 132 | |
More Information
Set Membership: |
|

HNRNPA2B1
Genomic Map