Assay Details
| Gene Symbol: PKNOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144108.3 | NP_001137580.1 | 6 - 7 | 810 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PKNOX2| Gene Symbol: PKNOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144108.3 | NP_001137580.1 | 6 - 7 | 810 | 57 | |
Set Membership: |
|