Assay Details
| Gene Symbol: STK38L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101092.2 | NP_001094562.1 | 6 - 7 | 514 | 126 | |
| GenBank mRNA | BC151309.1 | - | 6 - 7 | 515 | 126 | |
More Information
Set Membership: |
|

STK38L
Genomic Map