Assay Details
| Gene Symbol: PPP6C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130750.1 | NP_001124222.1 | 3 - 4 | 299 | 101 | |
| GenBank mRNA | BC112686.1 | - | 3 - 4 | 299 | 101 | |
More Information
Set Membership: |
|

PPP6C
Genomic Map