Assay Details
| Gene Symbol: MATR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102486.2 | NP_001095956.1 | 2 - 3 | 1153 | 84 | |
| GenBank mRNA | BC134450.1 | - | 2 - 3 | 1154 | 84 | |
More Information
Set Membership: |
|

MATR3
Genomic Map