Assay Details
| Gene Symbol: CKS1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113311.1 | NP_001106782.1 | 2 - 3 | 199 | 62 | |
| GenBank mRNA | BC120301.1 | - | 2 - 3 | 199 | 62 | |
More Information
Set Membership: |
|

CKS1B
Genomic Map