Assay Details
| Gene Symbol: IGFBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174555.1 | NP_776980.1 | 3 - 4 | 788 | 54 | |
| GenBank mRNA | AF074854.1 | - | 3 - 4 | 788 | 54 | |
More Information
Set Membership: |
|

IGFBP2
Genomic Map