Assay Details
| Gene Symbol: POU6F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102046.1 | NP_001095516.1 | 7 - 8 | 1491 | 83 | |
| GenBank mRNA | BC150031.1 | - | 7 - 8 | 1491 | 83 | |
More Information
Set Membership: |
|

POU6F2
Genomic Map