Assay Details
| Gene Symbol: DYRK1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081515.1 | NP_001074984.1 | 8 - 9 | 1335 | 157 | |
| GenBank mRNA | BC133387.1 | - | 8 - 9 | 1335 | 157 | |
More Information
Set Membership: |
|

DYRK1B
Genomic Map