Assay Details
| Gene Symbol: MTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206425.2 | NP_001193354.1 | 4 - 5 | 279 | 98 | |
| GenBank mRNA | BC148078.1 | - | 4 - 5 | 282 | 98 | |
More Information
Set Membership: |
|

MTM1
Genomic Map