Assay Details
| Gene Symbol: FAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077020.1 | NP_001070488.1 | 4 - 5 | 356 | 152 | |
| GenBank mRNA | BC105572.1 | - | 4 - 5 | 372 | 152 | |
More Information
Set Membership: |
|

FAF2
Genomic Map