Assay Details
| Gene Symbol: CALU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034665.2 | NP_001029837.1 | 3 - 4 | 484 | 97 | |
| GenBank mRNA | BC102361.1 | - | 3 - 4 | 485 | 97 | |
More Information
Set Membership: |
|

CALU
Genomic Map