Assay Details
| Gene Symbol: SLC25A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174658.2 | NP_777083.1 | 1 - 2 | 120 | 66 | |
| GenBank mRNA | BC102994.1 | - | 1 - 2 | 217 | 66 | |
| M24102.1 | - | 1 - 2 | 122 | 66 | ||
More Information
Set Membership: |
|

SLC25A4
Genomic Map