Assay Details
| Gene Symbol: KCNMB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001141.1 | NP_001001141.1 | 2 - 3 | 240 | 62 | |
| GenBank mRNA | BC147983.1 | - | 1 - 2 | 165 | 62 | |
| L26101.2 | - | 2 - 3 | 240 | 62 | ||
More Information
Set Membership: |
|

KCNMB1
Genomic Map