Assay Details
| Gene Symbol: MYDGF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001164.1 | NP_001001164.1 | 1 - 2 | 190 | 80 | |
| GenBank mRNA | AY581200.1 | - | 1 - 2 | 190 | 80 | |
| BC102581.1 | - | 1 - 2 | 207 | 80 | ||
More Information
Set Membership: |
|

MYDGF
Genomic Map