Assay Details
| Gene Symbol: ATG4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001170.1 | NP_001001170.1 | 5 - 6 | 457 | 64 | |
| GenBank mRNA | AY570550.1 | - | 5 - 6 | 457 | 64 | |
| AY570551.1 | - | 5 - 6 | 457 | 64 | ||
| BC102782.1 | - | 6 - 7 | 742 | 64 | ||
More Information
Set Membership: |
|

ATG4B
Genomic Map