Assay Details
| Gene Symbol: PPIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001597.1 | NP_001001597.1 | 3 - 4 | 316 | 57 | |
| GenBank mRNA | AY594335.1 | - | 3 - 4 | 316 | 57 | |
More Information
Set Membership: |
|

PPIF
Genomic Map