Assay Details
| Gene Symbol: PEG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002887.3 | NP_001002887.3 | 7 - 8 | 1103 | 69 | |
| GenBank mRNA | AY427787.1 | - | 1035 | 69 | ||
More Information
Set Membership: |
|

PEG3
Genomic Map