Assay Details
| Gene Symbol: PGGT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015560.1 | NP_001015560.1 | 1 - 2 | 150 | 68 | |
| GenBank mRNA | BT020634.1 | - | 1 - 2 | 150 | 68 | |
More Information
Set Membership: |
|

PGGT1B
Genomic Map