Assay Details
| Gene Symbol: CKB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015613.1 | NP_001015613.1 | 3 - 4 | 395 | 58 | |
| GenBank mRNA | BC102333.1 | - | 3 - 4 | 434 | 58 | |
| BT020708.1 | - | 3 - 4 | 395 | 58 | ||
More Information
Set Membership: |
|

CKB
Genomic Map