Assay Details
| Gene Symbol: GAPDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034034.2 | NP_001029206.1 | 3 - 4 | 187 | 66 | |
| GenBank mRNA | AB098979.1 | - | 3 - 4 | 175 | 66 | |
| BC102589.1 | - | 3 - 4 | 188 | 66 | ||
| DQ403066.1 | - | 1 - 2 | 41 | 66 | ||
| U85042.1 | - | 1 - 2 | 86 | 66 | ||
More Information
Set Membership: |
|

GAPDH
Genomic Map