Assay Details
| Gene Symbol: PCNA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034494.1 | NP_001029666.1 | 1 - 2 | 343 | 62 | |
| GenBank mRNA | BC103068.1 | - | 1 - 2 | 343 | 62 | |
More Information
Set Membership: |
|

PCNA
Genomic Map