Assay Details
| Gene Symbol: MOB3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034755.1 | NP_001029927.1 | 2 - 3 | 567 | 68 | |
| GenBank mRNA | BC149870.1 | - | 2 - 3 | 587 | 68 | |
| BT020665.1 | - | 2 - 3 | 567 | 68 | ||
More Information
Set Membership: |
|

MOB3C
Genomic Map