Assay Details
| Gene Symbol: NMI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035098.2 | NP_001030270.1 | 3 - 4 | 241 | 91 | |
| GenBank mRNA | BC102054.1 | - | 3 - 4 | 242 | 91 | |
| BC133621.1 | - | 3 - 4 | 312 | 91 | ||
More Information
Set Membership: |
|

NMI
Genomic Map