Assay Details
| Gene Symbol: CD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173899.2 | NP_776324.1 | 3 - 4 | 441 | 56 | |
| GenBank mRNA | BC113237.1 | - | 3 - 4 | 458 | 56 | |
| X53061.1 | - | 441 | 56 | |||
More Information
Set Membership: |
|

CD5
Genomic Map