Assay Details
| Gene Symbol: MIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173936.2 | NP_776361.1 | 3 - 4 | 448 | 65 | |
| GenBank mRNA | BC113340.1 | - | 1 - 2 | 1354 | 65 | |
| U51437.1 | - | 3 - 4 | 448 | 65 | ||
More Information
Set Membership: |
|

MIA
Genomic Map