Assay Details
| Gene Symbol: MX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173941.2 | NP_776366.1 | 6 - 7 | 997 | 66 | |
| GenBank mRNA | AF355147.1 | - | 6 - 7 | 997 | 66 | |
| BC149724.1 | - | 6 - 7 | 1012 | 66 | ||
More Information
Set Membership: |
|

MX2
Genomic Map