Assay Details
| Gene Symbol: SYP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173963.1 | NP_776388.1 | 5 - 6 | 638 | 59 | |
| GenBank mRNA | M22967.1 | - | 5 - 6 | 638 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SYP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173963.1 | NP_776388.1 | 5 - 6 | 638 | 59 | |
| GenBank mRNA | M22967.1 | - | 5 - 6 | 638 | 59 | |
Set Membership: |
|