Assay Details
| Gene Symbol: CCL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174006.2 | NP_776431.1 | 1 - 2 | 122 | 94 | |
| GenBank mRNA | BC142093.1 | - | 1 - 2 | 140 | 94 | |
| EU276059.1 | - | 1 - 2 | 96 | 94 | ||
| M84602.1 | - | 1 - 2 | 122 | 94 | ||
More Information
Set Membership: |
|

CCL2
Genomic Map