Assay Details
| Gene Symbol: CSF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174027.2 | NP_776452.1 | 3 - 4 | 339 | 71 | |
| GenBank mRNA | U22385.1 | - | 3 - 4 | 339 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CSF2| Gene Symbol: CSF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174027.2 | NP_776452.1 | 3 - 4 | 339 | 71 | |
| GenBank mRNA | U22385.1 | - | 3 - 4 | 339 | 71 | |
Set Membership: |
|