Assay Details
| Gene Symbol: FMOD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174058.2 | NP_776483.1 | 2 - 3 | 1031 | 80 | |
| GenBank mRNA | BC104615.1 | - | 2 - 3 | 1032 | 80 | |
| BT030742.1 | - | 2 - 3 | 1245 | 80 | ||
| X16485.1 | - | 1031 | 80 | |||
More Information
Set Membership: |
|

FMOD
Genomic Map