Assay Details
| Gene Symbol: HAS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174079.3 | NP_776504.2 | 3 - 4 | 1269 | 119 | |
| GenBank mRNA | AJ004951.1 | - | 3 - 4 | 1270 | 119 | |
More Information
Set Membership: |
|

HAS2
Genomic Map