Assay Details
| Gene Symbol: IL1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174092.1 | NP_776517.1 | 3 - 4 | 150 | 117 | |
| GenBank mRNA | M36182.1 | - | 3 - 4 | 127 | 117 | |
| M37210.1 | - | 3 - 4 | 150 | 117 | ||
| X12497.1 | - | 1 | 127 | 117 | ||
More Information
Set Membership: |
|

IL1A
Genomic Map