Assay Details
| Gene Symbol: MMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174112.1 | NP_776537.1 | 7 - 8 | 1048 | 74 | |
| GenBank mRNA | X58256.1 | - | 1048 | 74 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MMP1| Gene Symbol: MMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174112.1 | NP_776537.1 | 7 - 8 | 1048 | 74 | |
| GenBank mRNA | X58256.1 | - | 1048 | 74 | ||
Set Membership: |
|