Assay Details
| Gene Symbol: SERPINE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174137.2 | NP_776562.1 | 5 - 6 | 1018 | 59 | |
| GenBank mRNA | BC103451.1 | - | 5 - 6 | 1042 | 59 | |
| BT025406.1 | - | 5 - 6 | 1016 | 59 | ||
| KR006346.1 | - | 4 - 5 | 897 | 59 | ||
| X16383.1 | - | 1018 | 59 | |||
More Information
Set Membership: |
|

SERPINE1
Genomic Map