Assay Details
| Gene Symbol: DAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174162.1 | NP_776587.1 | 1 - 2 | 279 | 94 | |
| GenBank mRNA | AB009079.1 | - | 1 - 2 | 279 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DAG1| Gene Symbol: DAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174162.1 | NP_776587.1 | 1 - 2 | 279 | 94 | |
| GenBank mRNA | AB009079.1 | - | 1 - 2 | 279 | 94 | |
Set Membership: |
|