Assay Details
| Gene Symbol: STAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174189.3 | NP_776614.2 | 4 - 5 | 596 | 69 | |
| GenBank mRNA | BC110213.1 | - | 4 - 5 | 597 | 69 | |
| Y17259.1 | - | 69 | ||||
More Information
Set Membership: |
|

STAR
Genomic Map