Assay Details
| Gene Symbol: CHRNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174273.2 | NP_776698.1 | 9 - 10 | 1078 | 107 | |
| GenBank mRNA | BC149847.1 | - | 7 - 8 | 2139 | 107 | |
| M28307.1 | - | 9 - 10 | 1078 | 107 | ||
More Information
Set Membership: |
|

CHRNG
Genomic Map