Assay Details
| Gene Symbol: GNA14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174323.2 | NP_776748.1 | 4 - 5 | 1172 | 63 | |
| GenBank mRNA | D90335.1 | - | 4 - 5 | 1172 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GNA14| Gene Symbol: GNA14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174323.2 | NP_776748.1 | 4 - 5 | 1172 | 63 | |
| GenBank mRNA | D90335.1 | - | 4 - 5 | 1172 | 63 | |
Set Membership: |
|